Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD45 alpha Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI1C9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270559AF700

Catalog No. N270559A700


Add to cart

Description

Description

GADD45 alpha Monoclonal antibody specifically detects GADD45 alpha in Human samples. It is validated for Western Blot.
Specifications

Specifications

GADD45 alpha
OTI1C9
Western Blot
Affinity Purified, Conjugated
GADD45A
Full length recombinant protein of human GADD45 alpha (NP_001915) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Cancer
Primary
1647.0
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
DDIT-1, DDIT1growth arrest and DNA damage-inducible protein GADD45 alpha, DNA damage-inducible transcript 1 protein, DNA damage-inducible transcript-1, GADD45DNA-damage-inducible transcript 1, growth arrest and DNA-damage-inducible 45 alpha, growth arrest and DNA-damage-inducible, alpha
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit