Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GADD45G Rabbit anti-Human, DyLight 755, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP300285IR
Description
GADD45G Polyclonal antibody specifically detects GADD45G in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
GADD45G | |
DyLight 755 | |
50mM Sodium Borate | |
CR6DNA damage-inducible transcript 2 protein, Cytokine-responsive protein CR6, DDIT-2, DDIT2GADD45-gamma, growth arrest and DNA damage-inducible protein GADD45 gamma, growth arrest and DNA-damage-inducible, gamma, GRP1717 kD | |
Produced in rabbits immunized with purified, recombinant GADD45G (O95257; Met1-Glu159). | |
Affinity purified | |
RUO | |
Store at 4°C in the dark. | |
Polyclonal | |
Human |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified | |
Rabbit | |
IgG | |
0.1 mL | |
Apoptosis, Cancer | |
Primary | |
10912 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title