Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD45GIP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549337

Catalog No. PA549337

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a nuclear-localized protein that may be induced by p53 and regulates the cell cycle by inhibiting G1 to S phase progression. The encoded protein may interact with other cell cycle regulators. [provided by RefSeq, Aug 2012].
Specifications

Specifications

GADD45GIP1
Polyclonal
PBS with 0.09% sodium azide
Q8TAE8
Growth arrest and DNA damage-inducible proteins-interacting protein 1, 39S ribosomal protein L59, mitochondrial, MRP-L59, CKII beta-associating protein, CR6-interacting factor 1, CRIF1, Papillomavirus L2-interacting nuclear protein 1, PLINP, PLINP-1, p53-responsive gene 6 protein, GADD45GIP1, MRPL59, PLINP1, PRG6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
90480
Flow Cytometry, Western Blot
Unconjugated
GADD45GIP1
Liquid
GADD45GIP1
KLH conjugated synthetic peptide between 13-45 amino acids from the N-terminal region of human GADD45GIP1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit