Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium foil, 0.62mm (0.024in) thick, 99.9% (REO), Thermo Scientific™

Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
7440-54-2
Gd
157.25
MFCD00011022
UIWYJDYFSGRHKR-UHFFFAOYSA-N
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
23982
CHEBI:33375
gadolinium
[Gd]
7440-54-2
Gd
157.25
MFCD00011022
UIWYJDYFSGRHKR-UHFFFAOYSA-N
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
23982
CHEBI:33375
gadolinium
[Gd]
  • Packaged under argon
Weight ≈3.1g/25x25mm
Quantity 50 x 50 mm
Physical Form Foil
Percent Purity 99.9%
Chemical Name or Material Gadolinium foil, 0.62mm (0.024 in.) thick
Assay (REO)

EINECSNumber : 231-162-2

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

RUO – Research Use Only