Alfa Aesar™ Gadolinium(III) bromide hydrate, REacton™, 99.99% (REO)

Manufacturer: Alfa Aesar™ 1320822

Quantity 100g

 View more versions of this product

Catalog No. AA1320822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 30010-20-9
    Formula Weight 396.96 (Anhydrous)
    MDL Number MFCD00049741
    Chemical Name or Material Gadolinium(III) bromide hydrate
    Grade Reacton™

    Specifications

    Formula Weight 396.96 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00049741
    Quantity 100g
    TSCA TSCA
    Health Hazard 1 Warning
    Physical Form Crystalline
    Sensitivity Hygroscopic
    CAS 30010-20-9
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 186°C
    Chemical Name or Material Gadolinium(III) bromide hydrate
    Assay 99.99%, (REO)
    Grade Reacton™

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit