Alfa Aesar™ Gadolinium(III) carbonate hydrate, REacton™, 99.99% (REO)

Manufacturer: Alfa Aesar™ 1320722

Quantity 100g

 View more versions of this product

Catalog No. AA1320722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38245-36-2
    Formula Weight 494.53 (Anhydrous)
    MDL Number MFCD00150240
    Chemical Name or Material Gadolinium(III) carbonate hydrate
    Grade Reacton™

    Specifications

    Formula Weight 494.53 (Anhydrous)
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00150240
    CAS 38245-36-2
    Quantity 100g
    Chemical Name or Material Gadolinium(III) carbonate hydrate
    Assay 99.99%, (REO)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit