Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) chloride, 99.9%, (trace metal basis), anhydrous, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
10138-52-0
Cl3Gd
263.61
MFCD00011024
MEANOSLIBWSCIT-UHFFFAOYSA-K
gadolinium chloride,gadolinium trichloride,gadolinium chloride gdcl3,gdcl3,gadolinium iii chloride,gadolinium 3+ chloride,wln: gd g3,gadolinium iii chloride, ultra dry,gadolinium iii chloride, anhydrous, ?gdcl3
61486
CHEBI:37288
trichlorogadolinium
Cl[Gd](Cl)Cl
10138-52-0
Cl3Gd
263.61
MFCD00011024
MEANOSLIBWSCIT-UHFFFAOYSA-K
gadolinium chloride,gadolinium trichloride,gadolinium chloride gdcl3,gdcl3,gadolinium iii chloride,gadolinium 3+ chloride,wln: gd g3,gadolinium iii chloride, ultra dry,gadolinium iii chloride, anhydrous, ?gdcl3
61486
CHEBI:37288
trichlorogadolinium
Cl[Gd](Cl)Cl
for ultra trace analysis
Assay Percent Range 58.2 to 61.1% (Gd) (Complexometry)
Linear Formula GdCl3
Solubility Information Solubility in water: soluble
Formula Weight 263.61
Physical Form Powder
Percent Purity 99.90%
Grade Trace Metal Basis
Packaging Glass bottle
Total Trace Metal Impurities 1000ppm max.
Color White
Quantity 25g
Chemical Name or Material Gadolinium(III) chloride
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-386-6