Alfa Aesar™ Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO)

Manufacturer: Alfa Aesar™ 1128722

Quantity 100g

 View more versions of this product

Catalog No. AA1128722

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 13450-84-5
    Formula Weight 371.70 (263.61 Anhydrous)
    MDL Number MFCD00149732
    Merck Index 14,4326
    Chemical Name or Material Gadolinium(III) chloride hexahydrate
    Grade Reacton™

    Specifications

    Formula Weight 371.70 (263.61 Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Odor Characteristic
    Physical Form Crystalline Aggregates
    MDL Number MFCD00149732
    CAS 13450-84-5
    Merck Index 14,4326
    Color White
    Quantity 100g
    Health Hazard 1 Warning
    Density 2.424g/mL
    TSCA TSCA
    Sensitivity Hygroscopic
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    RTECS Number LW4050000
    Chemical Name or Material Gadolinium(III) chloride hexahydrate
    Assay 99.99%, (REO)
    Grade Reacton™

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit