Learn More
Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO)
Description
Gadolinium(III) chloride hexahydrate is employed in synthesis and NMR studies of the paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes with possible use as contrast agents for magnetic resonance imaging..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline Aggregates |
| Quantity | 5 g |
| Merck Index | 14,4326 |
| Solubility Information | soluble in water. |
| IUPAC Name | gadolinium(3+);trichloride;hexahydrate |
| Formula Weight | 371.70 (263.61 Anhydrous) |
| Percent Purity | 99.99% |
| Grade | Reacton™ |
| Odor | Characteristic |
| Show More |
Safety and Handling
| Hazard Category | H315-H319 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| EINECSNumber | 233-386-6 |
| RTECSNumber | LW4050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.