Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) fluoride 99.9%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Plastic bottle
13765-26-9
F3 Gd
MFCD00016113
13765-26-9
F3 Gd
MFCD00016113
Assay Percent Range 99.9%
Linear Formula GdF3
Formula Weight 214.24
Physical Form Powder
Percent Purity ≥99.85% (ex fluor)
Packaging Plastic bottle
Color White
Quantity 5g
Chemical Name or Material Gadolinium(III) fluoride

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with acids liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger