Alfa Aesar™ Gadolinium(III) isopropoxide, 99.9% (metals basis), 5% w/v in toluene/isopropanol (3:2)

Precursor for Gd2O3 films using Sol-Gel process

Manufacturer: Alfa Aesar™ 42277AE

Quantity 100mL

 View more versions of this product

Catalog No. AA42277AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14532-05-9
    Linear Formula Gd[OCH(CH3)2]3
    MDL Number MFCD00144381
    UN Number UN1993
    Chemical Name or Material Gadolinium(III) isopropoxide

    Specifications

    Health Hazard 2 GHS H Statement
    H225-H304-H361-H373-H315-H319-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P301+P330+P331-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Immediately call a POISON CENTER or doctor/physician.
    Linear Formula Gd[OCH(CH3)2]3
    Odor Aromatic
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Physical Form Liquid
    UN Number UN1993
    Health Hazard 1 Danger
    Color Pale yellow
    Sensitivity Moisture Sensitive
    Quantity 100mL
    MDL Number MFCD00144381
    CAS 14532-05-9
    Concentration 5% w/v in toluene/isopropanol (3:2)
    Chemical Name or Material Gadolinium(III) isopropoxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H225-H304-H361-H373-H315-H319-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P280h-P305+P351+P338-P301+P330+P331-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit