Alfa Aesar™ Gadolinium(III) isopropoxide, 99% (metals basis)

Sol-Gel synthesis of gadolinium compounds

Manufacturer: Alfa Aesar™ 4137706

Quantity 5g

 View more versions of this product

Catalog No. AA4137706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14532-05-9
    Linear Formula Gd[OCH(CH3)2]3
    Formula Weight 334.51
    MDL Number MFCD00144381
    UN Number UN2925
    Chemical Name or Material Gadolinium(III) isopropoxide

    Specifications

    Formula Weight 334.51
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Physical Form Powder
    Linear Formula Gd[OCH(CH3)2]3
    MDL Number MFCD00144381
    Solubility Information Soluble in anhydrous 2-propanol, toluene
    Health Hazard 1 Danger
    Color Off-white
    Sensitivity Air & Moisture Sensitive
    CAS 14532-05-9
    UN Number UN2925
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Health Hazard 2 GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.
    Quantity 5g
    Chemical Name or Material Gadolinium(III) isopropoxide
    Assay 99%, (metals basis)

    GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit