Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Gadolinium powder, -40 mesh, 99% (REO)
Click to view available options
Quantity:
2 g
10 g
50 g
Description
- Packaged under argon
Specifications
Quantity | 2 g |
UN Number | UN3178 |
Mesh Size | −40 Mesh |
Formula Weight | 157.25 |
Physical Form | Powder |
Percent Purity | 99% |
Color | Silver |
Chemical Name or Material | Gadolinium powder |
Assay | (REO) |
Safety and Handling
GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber : 231-162-2
RTECSNumber : LW3850000
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures