Alfa Aesar™ Gadolinium silicide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 3635609

Quantity 10g

 View more versions of this product

Catalog No. AA3635609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12134-75-7
    Formula Weight 213.42
    MDL Number MFCD00144382
    Chemical Name or Material Gadolinium silicide

    Specifications

    Formula Weight 213.42
    Odor Odorless
    Physical Form Pieces
    MDL Number MFCD00144382
    TSCA TSCA
    Particle Size 6mm & down
    Quantity 10g
    CAS 12134-75-7
    Chemical Name or Material Gadolinium silicide
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit