Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAGE1 Monoclonal Antibody (4G6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524395
Description
Peptide Sequence: MSWRGRSTYY WPRPRRYVQP PEMIGPMRPE QFSDEVEPAT PEEGEPATQR QDPAAAQEGE DEGASAGQGP KPEADSQEQG HPQTGCECED GPDGQEMDPP NPEEVKTPEE EMRSHYVAQT GILWLLMNNC FLNLSPRKP
GAGE1 Monoclonal antibody specifically detects GAGE1 in Human samples. It is validated for ELISA, Western Blot.Specifications
GAGE1 | |
Monoclonal | |
Unconjugated | |
GAGE1 | |
Antigen MZ2-F; Cancer/testis antigen 4.1; cancer/testis antigen 4.5; cancer/testis antigen family 4, member 1; cancer/testis antigen family 4, member 5; CT4.1; CT4.5; G antigen 1; G antigen 5; GAGE1; GAGE-1; GAGE5; GAGE-5; MZ2-F antigen | |
Mouse | |
Protein A | |
RUO | |
2543 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
4G6 | |
PBS with no preservative; pH 7.4 | |
0 | |
GAGE1 | |
GAGE1 (NP_001459.2, 1 a.a. ∽ 139 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction