Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAGE3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549017

Catalog No. PA549017


Add to cart

Description

Description

This gene belongs to a family of genes that are expressed in a variety of tumors but not in normal tissues, except for the testis. The sequences of the family members are highly related but differ by scattered nucleotide substitutions. The antigenic peptide YYWPRPRRY, which is also encoded by several other family members, is recognized by autologous cytolytic T lymphocytes. [provided by RefSeq, Jul 2008]
Specifications

Specifications

GAGE3
Polyclonal
PBS with 0.09% sodium azide
Q13067
G antigen 3, GAGE-3, Cancer/testis antigen 43, CT43, GAGE3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2575
Western Blot
Unconjugated
GAGE3
Liquid
GAGE3
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human GAGE3.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit