Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAK Mouse anti-Human, Mouse, Rat, Clone: 1C2, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | GAK |
---|---|
Clone | 1C2 |
Dilution | Western Blot 1 ug/ml |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP197511
|
Novus Biologicals
NBP197511 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
GAK Monoclonal specifically detects GAK in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GAK | |
Western Blot 1 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
2580 | |
Recombinant rat GAK. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
1C2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication | |
cyclin G associated kinase, cyclin-G-associated kinase, EC 2.7.11.1, FLJ16629, FLJ40395, MGC99654 | |
GAK | |
IgG1 | |
Protein A purified | |
Recognizes Rat GAK. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title