Learn More
GAL-4 Activation Domain Mouse anti-Yeast, Clone: 5C8-5, Exalpha

Mouse Monoclonal Antibody
Supplier: Vector Laboratories Z145M

Description
Recognizes GAL4 derivatives containing the C-terminal activation domain.
GAL-4 Activation Domain Monoclonal antibody specifically detects GAL-4 Activation Domain in Yeast samples. It is validated for Immunoprecipitation, Western BlotSpecifications
GAL-4 Activation Domain | |
Monoclonal | |
0.5-2 mg/mL | |
PBS with 0.08% sodium azide; pH | |
P04386 | |
Mouse | |
Protein A/G | |
RUO | |
855828 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot | |
5C8-5 | |
Unconjugated | |
GAL4 | |
antigen NY-CO-27; GAL4; Gal-4; GAL81; galactose-responsive transcription factor GAL4; galectin 4; galectin-4; L-36 lactose-binding protein; L36LBP; Lactose-binding lectin 4; lectin, galactoside binding soluble 4; lectin, galactoside-binding, soluble, 4; LGALS4; Regulatory protein GAL4; YPL248C | |
Hybridoma produced by the fusion of splenocytes from mice immunized with full length GAL4 expressed in E. Coli and mouse myeloma cells. | |
100 μg | |
Primary | |
Yeast | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.