Learn More
GAL3ST1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 185031AP
Catalog No. 50-173-2170
Add to cart
Description
Description
Catalyzes the sulfation of membrane glycolipids. Seems to prefer beta-glycosides at the non-reducing termini of sugar chains attached to a lipid moiety. Catalyzes the synthesis of galactosylceramide sulfate (sulfatide), a major lipid component of the myelin sheath and of monogalactosylalkylacylglycerol sulfate (seminolipid), present in spermatocytes. Also acts on lactosylceramide, galactosyl 1-alkyl-2-sn-glycerol and galactosyl diacylglycerol (in vitro).
Specifications
Specifications
GAL3ST1
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99999, Q9JHE4
Cerebroside sulfotransferase, CST, GAL3ST1, GalCer sulfotransferase
Rabbit
IgG
150 μL
-20°C
Primary
53897, 683713, 9514
Western Blot
Unconjugated
GAL3ST1
Liquid
Gal3st1
GAL3ST1 Fusion Protein Ag13353
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat