Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAL3ST4 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17932620UL

 View more versions of this product

Catalog No. NBP17932620


Add to cart

Description

Description

GAL3ST4 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

GAL3ST4
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
GAL3ST4
The immunogen for this antibody is Gal3st4. Peptide sequence RPLQFGSAKVLGYVLQSGLNQKDKEECERLATPELQYKDKLDAKQFPPTV.
Immunogen affinity purified
RUO
Primary
79690
Western Blot
Western Blot 1:1000
NP_001028588
EC 2.8.2.-, GAL3ST-4, galactose-3-O-sulfotransferase 44,Beta-galactose-3-O-sulfotransferase 4, Gal-beta-1,3-GalNAc 3'-sulfotransferase, galbeta1-3GalNAc 3'-sulfotransferase
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit