Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galacto-Star™ β-Galactosidase Reporter Gene Assay System for Mammalian Cells Non-distribution product as customer accommodation.

The Galacto-Star™ One-Step β-Galactosidase Reporter Gene Assay System chemiluminescent reporter gene assay system enables rapid, sensitive detection of β-galactosidase in mammalian cell lysates. Sensitive The chemiluminescent assay exhibits over three o

$327.00 - $716.00

Specifications

Product Type Reporter Gene Assays
For Use With (Application) Detection of β-galactosidase in mammalian cell lysates
For Use With (Equipment) Luminometer or scintillation counter
Format 96-well plate
Detection Method Enhanced Chemiluminescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
T1012 Non-distribution product as customer accommodation. View Documents Invitrogen™
T1012
600 assays Each for $327.00
Add to cart
 
T1014 Non-distribution product as customer accommodation. View Documents Invitrogen™
T1014
1,800 assays Each for $716.00
Add to cart
 
Description

Description

  • Glow assay kinetics provides a “window” during which measurements may be performed, facilitating HTS applications where assay automation is used
  • Can be used with either mammalian or yeast model systems, providing flexibility in choice of model systems
  • Wide dynamic range of β-galactosidase assay lets the user measure enzyme level accurately from femtogram to nanogram range
  • Assay sensitivity is 100- to 1,000-fold better than either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assays
  • Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions
  • Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes
  • Assay can be completed in about one hour, providing fast assay turnaround
Sensitive
The chemiluminescent assay exhibits over three orders of magnitude greater sensitivity than colorimetric β-galactosidase assays and can be completed in a fraction of the time required for assays for chloramphenicol acetyltransferase (CAT) and ELISA-based reporter assays.
 
Versatile
The Galacto-Star system includes Galacton-Star chemiluminescent substrate for β-galactosidase and is suited for measurement in both luminometers and scintillation counters. Automatic injection is not required.
 
Long-Lasting Signal
Light emission from the Galacto-Star assay reaches maximum in 60 to 90 minutes and remains constant for nearly one hour. Cell lysate is mixed with reaction buffer containing Galacton-Star substrate and Sapphire-II™ luminescence enhancer. After a 60-90 minute incubation at room temperature, the signal is measured in a luminometer or scintillation counter.
 
Specifications
  • Assay Type: β-Gal (lacZ) Assay
  • Compatible Cells: Mammalian Cells
  • Detection Method: Chemiluminescent
  • For Use With Equipment: Luminometer (Microplate)
  • Form: Liquid
  • Format: 96-well plate
  • High Throughput Compatibility: High Throughput-Compatible
  • Readout: End Point
  • Sample Type (Specific): Cell Cultures
  • Substrate: Galacton-Star
  • Substrate Properties: Chemical Substrate
  • Substrate Type: Beta-Gal Substrate
  • Target Enzyme: Beta-Galactosidase
Specifications

Specifications

Reporter Gene Assays
Luminometer or scintillation counter
Enhanced Chemiluminescence
Store at 2° to 8°C
Detection of β-galactosidase in mammalian cell lysates
96-well plate
Lysis solution, substrate, enhancer, and required buffer

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit