Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galacto-Star™ Reaction Buffer Diluent with Galacton-Star™ Substrate Non-distribution product as customer accommodation.

Substrate solution and substrate dilution buffer for detection of E.coli β-galactosidase. Thermo Scientific Galacto-Star Reaction Buffer Diluent with Galacton-Star Substrate are for use with Galacto-Star One-Step β-Galactosidase Reporter Gene As

Manufacturer:  Invitrogen™ T1056

Catalog No. T1056


Add to cart

Includes: Concentrated substrate and dilution buffer

Description

Description

For use with Galacto-Star One-Step β-Galactosidase Reporter Gene Assay System.
Specifications

Specifications

Detection of β-galactosidase
Substrate and buffer diluent
1800 assays
96-well plate
Galacto-Star Reaction Buffer Diluent with Galacton-Star Substrate Reporter Gene Assays
Store at 2° to 8°C
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.