Learn More
BioVision IncSupplier Diversity Partner Galactose Colorimetric/Fluorometric Assay Kit: Galactose Assay Kit: High-Throughput Colorimetric & Fluorometric Assay to Measure Galactose levels from various Biological Samples within 40 min. Rapid, Convenient & Sensitive. 100 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K621100
Galactose (C₆H₁₂O₆ FW: 180.16) is one the major naturally occurring sugars, involved in many biological processes. BioVision’s Galactose Assay Kit provides a simple, convenient means for direct measurement of galactose levels in various biological samples (serum, plasma, other body fluids, food, growth media, etc.). In the assay kit, Galactose is specifically oxidized generating a product that produces color (O.D 570 nm) and fluorescence (Ex/Em 535/587 nm). Liquid samples can be tested directly without purification. The assay is fast, convenient and sensitive. It can also be used as a high throughput assay. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9801