Learn More
Alfa Aesar™ Galanin (1-16), Porcine, rat
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66716LB0
 View more versions of this product
Catalog No. AAJ66716LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1669.92
Porcine, Rat
Solid
RUO
Galanin (1-16), porcine, rat
HPLC
5mg
Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Ile
-30° to -10°C