Learn More
Alfa Aesar™ Galanin (2-11)
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66796MCR
 View more versions of this product
Catalog No. AAJ66796MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1136.30
1mg
Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-NH2
-30° to -10°C
HPLC
Solid
RUO
Galanin (2-11)