Learn More
Thermo Scientific Chemicals Galanin, Human
For cellular and molecular biology applications
$209.00 - $373.00
Specifications
Species | Human |
---|---|
Purification Method | HPLC |
Content And Storage | -30°C to -10°C |
Format | Powder |
Molecular Weight (g/mol) | 3157.41 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ66251EXD
|
Thermo Scientific Chemicals
J66251EXD |
0.5 mg |
Each for $209.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ66251MCR
|
Thermo Scientific Chemicals
J66251MCR |
1 mg |
Each for $373.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Human | |
-30°C to -10°C | |
3157.41 | |
RUO - research use only |
HPLC | |
Powder | |
Galanin, human | |
Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Val-Gly-Asn-His-Arg-Ser-Phe-Ser-Asp-Lys-Asn-Gly-Leu-Thr-Ser |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.