Learn More
Alfa Aesar™ Galanin, Human
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66251EXD
 View more versions of this product
Catalog No. AAJ66251EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
3157.41
Human
Powder
RUO
Galanin, human
HPLC
0.5mg
Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Val-Gly-Asn-His-Arg-Ser-Phe-Ser-Asp-Lys-Asn-Gly-Leu-Thr-Ser
-30° to -10°C