Learn More
Selleck Chemical LLCSupplier Diversity Partner Galanthamine HBr 500mg 1953-04-4  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S1339500MG
Galanthamine is an AChE inhibitor with IC50 of 0.35 μM, exhibits 50-fold selectivity against butyryl-cholinesterase.
Catalog No. 50-627-8