Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
α-GalCer:CD1d Complex Mouse anti-Mouse, PE, Clone: L363, BD Phrmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569977
Description
α-GalCer:CD1d Complex Monoclonal antibody specifically detects α-GalCer:CD1d Complex in Mouse samples. It is validated for Flow cytometry
Specifications
α-GalCer:CD1d Complex | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
12479 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2a κ |
Flow Cytometry | |
L363 | |
PE | |
α-GalCer:mCD1d; αGalCer-CD1d; αlpha-GalCer:CD1d; αlpha-GalCer:Mouse CD1d | |
α-GalCer:MouseCD1d1 complexes coupled to M. tuberculosis PPD | |
0.1 mg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction