Learn More
GALE Antibody (OTI1C4), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270793FR
Catalog No. NBP270793FR
Add to Cart
Description
Description
GALE Monoclonal specifically detects GALE in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
GALE
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
GALE
Full length recombinant protein of human GALE (NP_000394) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat, Canine, Primate, Monkey
IgG1
Western Blot, Immunohistochemistry
OTI1C4
Western Blot, Immunohistochemistry
EC 5.1.3, EC 5.1.3.2, FLJ95174, FLJ97302, Galactowaldenase, short chain dehydrogenase/reductase family 1E, member 1, UDP-, UDP galactose-4′-epimerase, UDP-galactose 4-epimerase, UDP-galactose-4-epimerase, UDP-glucose 4-epimerase
Mouse
Affinity Purified
Lipid and Metabolism
2582.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions