Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALE Mouse anti-Human, Rat, Canine, Monkey, Alexa Fluor™ 647, Clone: OTI1C4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270793AF647

Catalog No. N270793A647


Add to cart

Description

Description

GALE Monoclonal antibody specifically detects GALE in Human, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

GALE
OTI1C4
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
GALE
Full length recombinant protein of human GALE (NP_000394) produced in HEK293T cells.
Immunogen affinity purified
Lipid and Metabolism
Primary
2582.0
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
EC 5.1.3, EC 5.1.3.2, FLJ95174, FLJ97302, Galactowaldenase, short chain dehydrogenase/reductase family 1E, member 1, UDP-, UDP galactose-4′-epimerase, UDP-galactose 4-epimerase, UDP-galactose-4-epimerase, UDP-glucose 4-epimerase
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Canine, Human, Monkey, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit