Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-1 Mouse anti-Human, PE/Cy5.5, Clone: GAL1/1831, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254457PECY55

Catalog No. N254457PEC5


Add to cart

Description

Description

Galectin-1 Monoclonal antibody specifically detects Galectin-1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Galectin-1
GAL1/1831
Flow Cytometry
Conjugated, Purified
LGALS1
Recombinant human Galectin-1 protein fragment (aa12-108) (exact sequence is proprietary)
Protein A or G purified
Apoptosis, Biologically Active Proteins
Primary
3956.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1
Mouse
IgG1 Lambda
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit