Learn More
Galectin-1 Rat anti-Mouse, DyLight 405, Clone: RM0081-9J26, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP121721V
Catalog No. NBP121721V
Add to cart
Description
Description
Galectin-1 Monoclonal antibody specifically detects Galectin-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
Galectin-1
RM0081-9J26
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
LGALS1
Mouse recombinant Galectin-1
Protein G purified
Apoptosis, Biologically Active Proteins
Primary
3956
Mouse
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1
Rat
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Galectin-1.