Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-3 Mouse, Alexa Fluor 647, Clone: 5C21, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192690AF647

Catalog No. NBP192690AH


Add to cart

Description

Description

Galectin-3 Monoclonal antibody specifically detects Antigen in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blotting.
Specifications

Specifications

Galectin-3
5C21
Western Blot, Immunocytochemistry/Immunofluorescence
Affinity Purified
LGALS3
Full length recombinant human Galectin-3 expressed in and purified from E. coli. [UniProt# P17931]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3
Mouse
IgG1
0.1 ml
Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors
Primary
3958
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit