Learn More
Galectin-3 Mouse, Alexa Fluor 647, Clone: B2C10, BD Pharmingen™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565678
Catalog No. BDB565678
Add to cart
Description
Description
Galectin-3 Monoclonal antibody specifically detects Galectin-3 in Human, Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
Specifications
Galectin-3
B2C10
Alexa Fluor 647
Purified
Murine
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions, and unreacted Alexa Fluor™ 647 was removed.
Monoclonal
Flow Cytometry, Flow Cytometry (Intracellular Staining)
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
LGALS3; Gal-3; GAL3; GALIG; GALBP; CBP35; MAC-2; IgE-binding protein; L31
Recombinant Human Galectin-3
50μg
Cell Biology
Primary
Human, Murine
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.