Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-3 Mouse anti-Human, Mouse, Alexa Fluor™ 488, Clone: B2C10, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565677
Description
Galectin-3 Monoclonal antibody specifically detects Galectin-3 in Human,Mouse samples. It is validated for Flow Cytometry.Specifications
Galectin-3 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Galectin-3 | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
B2C10 | |
Alexa Fluor 488 | |
50 μg | |
Primary | |
Human, Mouse | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title