Learn More
Galectin-3 Mouse anti-Human, Mouse, PE-CF594, Clone: B2C10, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565682
Catalog No. BDB565682
Add to cart
Description
Description
Galectin-3 Monoclonal antibody specifically detects Galectin-3 in Human,Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Galectin-3
B2C10
PE-CF594
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Human, Mouse
Flow Cytometry
0.2 mg/mL
Conjugated, Purified
IgG1, κ
RUO
Primary
Galectin-3