Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$212.55 - $436.00

Specifications

Antigen Galectin-3
Applications Western Blot, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092943
View Documents
Novus Biologicals
NBP303252100UL
100 μL
Each for $436.00
Add to cart
 
NB092944
View Documents
Novus Biologicals
NBP30325220UL
20 μL
Each for $212.55
Add to cart
 
Description

Description

Galectin-3 Polyclonal specifically detects Galectin-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications

Specifications

Galectin-3
Polyclonal
Purified
RUO
Human, Mouse, Rat
Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3
Recombinant fusion protein containing a sequence corresponding to amino acids 1-1-250 of human Galectin-3 (NP_002297.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot, Immunofluorescence, KnockDown
Unconjugated
Rabbit
Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors
PBS with 50% glycerol, pH7.3.
3958
IgG
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit