Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-3BP/MAC-2BP/LGALS3BP Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI6B7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271125JF646

Catalog No. N271125J646


Add to cart

Description

Description

Galectin-3BP/MAC-2BP/LGALS3BP Monoclonal antibody specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

Galectin-3BP/MAC-2BP/LGALS3BP
OTI6B7
Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
LGALS3BP
Recombinant protein fragment corresponding to amino acids 19-300 of human Galectin-3BP/MAC-2BP/LGALS3BP (NP_005558) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K
Mouse
IgG2a
0.1 mL
Primary
3959.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit