Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-3BP/MAC-2BP/LGALS3BP Antibody (OTI6B7), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271125PECY7
Description
Galectin-3BP/MAC-2BP/LGALS3BP Monoclonal specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human samples. It is validated for Flow Cytometry.Specifications
Galectin-3BP/MAC-2BP/LGALS3BP | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
LGALS3BP | |
Recombinant protein fragment corresponding to amino acids 19-300 of human Galectin-3BP/MAC-2BP/LGALS3BP (NP_005558) produced in E.coli. | |
0.1 mL | |
3959.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI6B7 | |
Flow Cytometry | |
90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction