Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-3BP/MAC-2BP/LGALS3BP Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Galectin-3BP/MAC-2BP/LGALS3BP
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093095
View Documents
Novus Biologicals
NBP303669100UL
100 μg Each for $320.71
Add to cart
 
NB093096
View Documents
Novus Biologicals
NBP30366920UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Galectin-3BP/MAC-2BP/LGALS3BP Polyclonal antibody specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

Galectin-3BP/MAC-2BP/LGALS3BP
Unconjugated
Rabbit
Human, Mouse, Rat
90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K
Recombinant fusion protein containing a sequence corresponding to amino acids 306-585 of human Galectin-3BP/MAC-2BP/LGALS3BP (NP_005558.1).
Polyclonal
Affinity purified
Immunofluorescence, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
3959
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit