Learn More
Galectin-7 Rat anti-Mouse, DyLight 350, Clone: RM0083-8J52, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP121723UV
Catalog No. NBP121723UV
Add to cart
Description
Description
Galectin-7 Monoclonal antibody specifically detects Galectin-7 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Galectin-7
RM0083-8J52
Western Blot
Purified
LGALS7
Mouse recombinant Galectin-7
Protein G purified
Apoptosis, Cancer, Tumor Suppressors
Primary
3963
Mouse
Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Gal-7, GAL7, galectin 7, galectin-7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Galectin-7.