Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-7 Rat anti-Mouse, DyLight 405, Clone: RM0083-8J52, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP121723V
Description
Galectin-7 Monoclonal antibody specifically detects Galectin-7 in Mouse samples. It is validated for Western Blot.Specifications
Galectin-7 | |
RM0083-8J52 | |
Western Blot | |
Purified | |
LGALS7 | |
Mouse recombinant Galectin-7 | |
Protein G purified | |
Apoptosis, Cancer, Tumor Suppressors | |
Primary | |
3963 | |
Mouse |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Gal-7, GAL7, galectin 7, galectin-7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1 | |
Rat | |
IgG2 | |
0.1 ml | |
Store at 4C in the dark. | |
Monoclonal | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Galectin-7. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title