Learn More
Galectin 8 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA550965
Catalog No. PA550965
Add to cart
Description
Description
Galectin 8 Polyclonal Antibody detects endogenous levels of Galectin 8 protein.
Lectin with a marked preference for 3 ft.-O-sialylated and 3 ft.-O-sulfated glycans. {ECO:0000269; PubMed:21288902}.
Specifications
Specifications
Galectin 8
Polyclonal
Unconjugated
LGALS8
Liquid
LGALS8
Synthesized peptide derived from the Internal region of human Galectin-8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
O00214, Q9JL15
LGALS8, Galectin-8, Gal-8, Po66 carbohydrate-binding protein, Po66-CBP, Prostate carcinoma tumor antigen 1, PCTA-1
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3964, 56048