Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-8 Rabbit anti-Human, Mouse, Rat, Clone: JB85-35, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275501
Description
Galectin-8 Monoclonal specifically detects Galectin-8 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Galectin-8 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:400-1:800, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200 | |
Gal-8, galectin 8, galectin-8, galectin-8g, lectin, galactoside-binding, soluble, 8, PCTA1, PCTA-1Po66 carbohydrate-binding protein, Po66 carbohydrate binding protein, Po66-CBP, Prostate carcinoma tumor antigen 1 | |
Rabbit | |
40 kDa | |
100 μL | |
3964.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
JB85-35 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
LGALS8 | |
Synthetic peptide within Human Galectin-8 aa 1-50/ 317. (SwissProt: O00214 Human; SwissProt: Q9JL15 Mouse; SwissProt: Q62665 Rat) | |
Protein A purified | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction