Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin 9 Mouse anti-Human, eFluor® 660, Clone: 9M1-3, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 50911641

 View more versions of this product

Catalog No. 50-112-4498


Add to cart

Description

Description

Description: This 9M1-3 monoclonal antibody reacts with human Galectin-9. Galectin-9 is one of fifteen members of the galectin family of glycan-binding proteins. It is expressed on activated T helper cells and Foxp3+ T regulatory cells and plays a critical role in the regulation of immune function. Interaction of Galectin-9 with its ligand, TIM3, induces apoptosis in Th1 and Th17, inhibits Th17 polarization and IFN gamma expression, and drives Treg expansion. Galectin-9 is also thought to contribute to the survival and expansion of some tumors by inhibiting the anti-tumor response and promoting immune evasion. Along with Galectin-1 and Galectin-3, high levels of Galectin-9 expression are associated with poor cancer prognosis, making these potential therapeutic targets. Applications Reported: This 9M1-3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 9M1-3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm).

Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 µm post-manufacturing filtered. The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin ft.s disease tissue and might participate in the interaction between the H&RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications

Specifications

Galectin 9
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
O00182
HUAT, LGALS9A, ecalectin, gal-9, galectin 9, galectin-9, tumor antigen HOM-HD-21, urate transporter/channel protein
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
9M1-3
eFluor™ 660
LGALS9
Liquid
LGALS9
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3965
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.