Learn More
Galectin-9 Mouse anti-Human, PE, Clone: 9M1-3, BD Pharmingen™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565892
 View more versions of this product
Catalog No. BDB565892
Add to cart
Description
Description
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
Specifications
Galectin-9
9M1-3
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
galectin 9; Gal-9; Ecalectin; HUAT; LGALS9; LEG9; Tumor antigen HOM-HD-21
Human Galectin-9 (AA 208-215) Peptide
25 Tests
Cell Biology
Primary
Human
Flow Cytometry, Flow Cytometry (Intracellular Staining)
PE
Purified
Mouse
Mouse IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed.
Monoclonal
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.