Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-9 Mouse anti-Human, PE, Clone: 9M1-3, BD Pharmingen™

Mouse Monoclonal Antibody

$150.00 - $299.00

Specifications

Antigen Galectin-9
Isotype Mouse IgG1, κ
Immunogen Human Galectin-9 (AA 208-215) Peptide
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Conjugate PE
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB565892
View Documents
BD Biosciences
565892
25 Tests Each for $150.00
Add to cart
 
BDB565890
View Documents
BD Biosciences
565890
100 Tests Each for $299.00
Add to cart
 
Description

Description

Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .

Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications

Specifications

Galectin-9
Human Galectin-9 (AA 208-215) Peptide
PE
Cell Biology
Murine
Purified
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions
Mouse IgG1, κ
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Flow Cytometry, Flow Cytometry (Intracellular Staining)
RUO
9M1-3
galectin 9; Gal-9; Ecalectin; HUAT; LGALS9; LEG9; Tumor antigen HOM-HD-21

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit