Learn More
Galectin 9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA550966
Catalog No. PA550966
Add to cart
Description
Description
Galectin 9 Polyclonal Antibody detects endogenous levels of Galectin 9 protein.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin ft.s disease tissue and might participate in the interaction between the H&RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
Galectin 9
Polyclonal
Unconjugated
LGALS9
Liquid
LGALS9
Synthesized peptide derived from the Internal region of human Galectin-9.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
O00182, O08573, P97840
LGALS9, Galectin-9, Gal-9, Ecalectin, Tumor antigen HOM-HD-21
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
16859, 25476, 3965