Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-9 Rat anti-Mouse, PE, Clone: RG9-35, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 566027
Description
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .Specifications
Galectin-9 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
RG9-35 | |
PE | |
Galectin9; Gal-9; Gal9; Lgals9; LEG9 | |
Mouse Galectin-9 Recombinant Protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?
Provide Content Correction